tert-Butyl N-(1,3-oxazol-4-yl)carbamate 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl oxazol-4-ylcarbamate

OR933138-1G
APOSOR933138-1GEA 81.2 EUR
APOSOR933138-1G
tert-Butyl N-(1,3-oxazol-4-yl)carbamate 95%
tert-Butyl N-(1,3-oxazol-4-yl)carbamate
Formel: C₈H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18533643
CAS-Nummer: 1314931-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR