Sodium glycochenodeoxycholate 95%

Lieferant: Apollo Scientific

OR933168-1G OR933168-25G OR933168-5G
APOSOR933168-1GEA 43.5 EUR
APOSOR933168-1G APOSOR933168-25G APOSOR933168-5G
Sodium glycochenodeoxycholate 95%
Sodium glycochenodeoxycholate
Formel: C₂₆H₄₂NNaO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00036743
CAS-Nummer: 16564-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR