Piperidine-1-sulphonyl chloride 97%
Lieferant: Apollo Scientific
OR933169-25G
APOSOR933169-25GEA
282
EUR
APOSOR933169-25G
Piperidine-1-sulphonyl chloride 97%
Piperidine-1-sulphonyl chloride
Formel:
C₅H₁₀ClNO₂S Molecular Weight: 183.65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03250329 CAS-Nummer: 35856-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...