4-Chloro-2-methyl-6,7-dihydro-5h-cyclopenta[d]pyrimidine 90%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-methyl-6,7-dihydro-5H-cyclopenta[d]pyrimidine

OR933192-1G
APOSOR933192-1GEA 243 EUR
APOSOR933192-1G
4-Chloro-2-methyl-6,7-dihydro-5h-cyclopenta[d]pyrimidine 90%
4-Chloro-2-methyl-6,7-dihydro-5h-cyclopenta[d]pyrimidine
Formel: C₈H₉ClN₂
Molecular Weight: 168,623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14648013
CAS-Nummer: 118802-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR