2-Nitro-5-(piperazin-1-yl)aniline

Lieferant: Apollo Scientific

OR933206-1G
APOSOR933206-1GEA 154 EUR
APOSOR933206-1G
2-Nitro-5-(piperazin-1-yl)aniline
2-Nitro-5-(piperazin-1-yl)aniline
Formel: C₁₀H₁₄N₄O₂
Molecular Weight: 222,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00545958
CAS-Nummer: 96103-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR