2-Hydroxy-4-methylbenzyl alcohol 95%

Lieferant: Apollo Scientific

OR933259-1G
APOSOR933259-1GEA 734 EUR
APOSOR933259-1G
2-Hydroxy-4-methylbenzyl alcohol 95%
2-Hydroxy-4-methylbenzyl alcohol
Formel: C₈H₁₀O₂
Molecular Weight: 138,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202811
CAS-Nummer: 7405-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR