N-Carbobenzoxy-DL-Norvalin 97%

Lieferant: Apollo Scientific

Synonyme: Z-DL-Nva-OH, N-Benzyloxycarbonyl-DL-norvaline, N-Cbz-DL-norvaline

OR933315-25G
APOSOR933315-25GEA 139 EUR
APOSOR933315-25G
N-Carbobenzoxy-DL-Norvalin 97%
N-Carbobenzoxy-DL-Norvalin
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020434
CAS-Nummer: 21691-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR