meso-1,4-Dichloro-2,3-butanediol 95%

Lieferant: Apollo Scientific

OR933328-5G
APOSOR933328-5GEA 377 EUR
APOSOR933328-5G
meso-1,4-Dichloro-2,3-butanediol 95%
meso-1,4-Dichloro-2,3-butanediol
Formel: C₄H₈Cl₂O₂
Molecular Weight: 159,01 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7268-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR