3-(4-Chloro-1H-pyrazol-1-yl)propanoic acid 95%

Lieferant: Apollo Scientific

OR933358-1G
APOSOR933358-1GEA 319 EUR
APOSOR933358-1G
3-(4-Chloro-1H-pyrazol-1-yl)propanoic acid 95%
3-(4-Chloro-1H-pyrazol-1-yl)propanoic acid
Formel: C₆H₇ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03419657
CAS-Nummer: 913839-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR