4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 95%

Lieferant: Apollo Scientific

Synonyme: 4,6-Dihydroxy-5-methylisophthalaldehyde

OR933368-1G
APOSOR933368-1GEA 101 EUR
APOSOR933368-1G
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 95%
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene
Formel: C₉H₈O₄
Molecular Weight: 180,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505923
CAS-Nummer: 22304-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR