Methyl 3-(benzyloxy)propanoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Benzyloxy-propionic acid methyl ester

OR933371-1G
APOSOR933371-1GEA 91.5 EUR
APOSOR933371-1G
Methyl 3-(benzyloxy)propanoate 98%
Methyl 3-(benzyloxy)propanoate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274890
CAS-Nummer: 4126-60-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR