4-Bromo-1-cyclopropyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR933399-1G
APOSOR933399-1GEA 55.1 EUR
APOSOR933399-1G
4-Bromo-1-cyclopropyl-1H-pyrazole 95%
4-Bromo-1-cyclopropyl-1H-pyrazole
Formel: C₆H₇BrN₂
Molecular Weight: 187,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18250639
CAS-Nummer: 1151802-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR