1-(4-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR9334-1G
APOSOR9334-1GEA 52.2 EUR
APOSOR9334-1G
1-(4-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₂H₁₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00227562
CAS-Nummer: 56617-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR