4-Butylpyridine 95%

Lieferant: Apollo Scientific

OR933401-1G
APOSOR933401-1GEA 574 EUR
APOSOR933401-1G
4-Butylpyridine 95%
4-Butylpyridine
Formel: C₉H₁₃N
Molecular Weight: 135,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129015
CAS-Nummer: 5335-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR