2-(Dimethylaminomethyl)-3-hydroxypyridine 95%

Lieferant: Apollo Scientific

Synonyme: 2-((Dimethylamino)methyl)pyridin-3-ol

OR933407-25G
APOSOR933407-25GEA 197 EUR
APOSOR933407-25G
2-(Dimethylaminomethyl)-3-hydroxypyridine 95%
2-(Dimethylaminomethyl)-3-hydroxypyridine
Formel: C₈H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00006347
CAS-Nummer: 2168-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR