(6-Methylpyridazin-3-yl)methanamine dihydrochloride

Lieferant: Apollo Scientific

OR933478-1G
APOSOR933478-1GEA 833 EUR
APOSOR933478-1G
(6-Methylpyridazin-3-yl)methanamine dihydrochloride
(6-Methylpyridazin-3-yl)methanamine dihydrochloride
Formel: C₆H₉N₃·2ClH
Molecular Weight: 196,078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28155130
CAS-Nummer: 1630907-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR