3-(4-Nitrophenyl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR9335-1G
OR9335-250MG
APOSOR9335-1GEA
65.4
EUR
APOSOR9335-1G
APOSOR9335-250MG
3-(4-Nitrophenyl)-1H-pyrazole
3-(4-Nitrophenyl)-1H-pyrazole
Formel:
C₉H₇N₃O₂ Schmelzpunkt: 194…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665859 CAS-Nummer: 20583-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...