2-(Azetidin-3-yl)acetonitrile hydrochloride

Lieferant: Apollo Scientific

OR933553-1G
APOSOR933553-1GEA 432 EUR
APOSOR933553-1G
2-(Azetidin-3-yl)acetonitrile hydrochloride
2-(Azetidin-3-yl)acetonitrile hydrochloride
Formel: C₅H₈N₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28501222
CAS-Nummer: 1423057-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR