Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Lieferant: Apollo Scientific
OR933912-1G
OR933912-250MG
OR933912-500MG
APOSOR933912-1GEA
1490
EUR
APOSOR933912-1G
APOSOR933912-250MG
APOSOR933912-500MG
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Formel:
C₇H₁₃NO₂·ClH Molecular Weight: 179.645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28014565 CAS-Nummer: 1630907-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...