Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR933912-1G OR933912-250MG OR933912-500MG
APOSOR933912-1GEA 1490 EUR
APOSOR933912-1G APOSOR933912-250MG APOSOR933912-500MG
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Formel: C₇H₁₃NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28014565
CAS-Nummer: 1630907-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR