4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Tert-butylcarbonylamino)phenylboronic acid, 4-Pivalamidobenzeneboronic acid

OR9346-1G
APOSOR9346-1GEA 372 EUR
APOSOR9346-1G
4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid 98%
4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221,061 g/mol
Schmelzpunkt: 240…243 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235071
CAS-Nummer: 182344-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR