2-Phenyl-1-(piperazin-1-yl)ethan-1-one hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 2-Phenyl-1-(piperazin-1-yl)ethanone hydrochloride

OR934729-1G OR934729-250MG
APOSOR934729-1GEA 132 EUR
APOSOR934729-1G APOSOR934729-250MG
2-Phenyl-1-(piperazin-1-yl)ethan-1-one hydrochloride 95%
2-Phenyl-1-(piperazin-1-yl)ethan-1-one hydrochloride
Formel: C₁₂H₁₆N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28040565
CAS-Nummer: 502653-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR