2-Norbornene-2-boronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (Bicyclo[2.2.1]hept-2-en-2-yl)boronic acid

OR9349-250MG
APOSOR9349-250MGEA 655 EUR
APOSOR9349-250MG
2-Norbornene-2-boronic acid 95%
2-Norbornene-2-boronic acid
Formel: C₇H₁₁BO₂
Molecular Weight: 137.97 g/mol
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07783853
CAS-Nummer: 871333-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR