(1S)-1,7,7-Trimethylnorborn-2-en-2-boronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: [(1S)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-en-2-yl]boronic acid

OR9350-250MG
APOSOR9350-250MGEA 466 EUR
APOSOR9350-250MG
(1S)-1,7,7-Trimethylnorborn-2-en-2-boronsäure 95%
(1S)-1,7,7-Trimethylnorborn-2-en-2-boronsäure
Formel: C₁₀H₁₇BO₂
Molecular Weight: 180,05 g/mol
Schmelzpunkt: 205…207 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08705231
CAS-Nummer: 871333-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR