5-Azaspiro[2.5]octan-8-one hydrochloride

Lieferant: Apollo Scientific

OR935112-1G
APOSOR935112-1GEA 2450 EUR
APOSOR935112-1G
5-Azaspiro[2.5]octan-8-one hydrochloride
5-Azaspiro[2.5]octan-8-one hydrochloride
Formel: C₇H₁₁NO·ClH
Molecular Weight: 161,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28501958
CAS-Nummer: 1780166-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR