5-Bromobenzo[d]isothiazol-3(2H)-one ≥98%
Lieferant: Apollo Scientific
OR935139-250MG
APOSOR935139-250MGEA
244
EUR
APOSOR935139-250MG
5-Bromobenzo[d]isothiazol-3(2H)-one ≥98%
5-Bromobenzo[d]isothiazol-3(2H)-one
Formel:
C₇H₄BrNOS Molecular Weight: 230,082 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28671026 CAS-Nummer: 186584-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...