6-Bromo-2-chloro-3-nitroquinoline ≥96%

Lieferant: Apollo Scientific

OR935238-250MG
APOSOR935238-250MGEA 126 EUR
APOSOR935238-250MG
6-Bromo-2-chloro-3-nitroquinoline ≥96%
6-Bromo-2-chloro-3-nitroquinoline
Formel: C₉H₄BrClN₂O₂
Molecular Weight: 287,497 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27928359
CAS-Nummer: 754990-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR