6-Methyl-2-azaspiro[3.3]heptan-6-ol hydrochloride

Lieferant: Apollo Scientific

OR935289-250MG
APOSOR935289-250MGEA 488 EUR
APOSOR935289-250MG
6-Methyl-2-azaspiro[3.3]heptan-6-ol hydrochloride
6-Methyl-2-azaspiro[3.3]heptan-6-ol hydrochloride
Formel: C₇H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28501954
CAS-Nummer: 1638765-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR