5-Oxocyclopenten-1-ylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Oxocyclopentene-1-boronic acid

OR9353-250MG
APOSOR9353-250MGEA 698 EUR
APOSOR9353-250MG
5-Oxocyclopenten-1-ylboronsäure 97%
5-Oxocyclopenten-1-ylboronic acid
Formel: C₅H₇BO₃
Molecular Weight: 125,92 g/mol
Schmelzpunkt: 106…108 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08056354
CAS-Nummer: 871329-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR