Methyl 2-{3-azaspiro[5.5]undecan-9-yl}acetate hydrochloride

Lieferant: Apollo Scientific

OR935882-1G
APOSOR935882-1GEA 1490 EUR
APOSOR935882-1G
Methyl 2-{3-azaspiro[5.5]undecan-9-yl}acetate hydrochloride
Methyl 2-{3-azaspiro[5.5]undecan-9-yl}acetate hydrochloride
Formel: C₁₃H₂₃NO₂·ClH
Molecular Weight: 261,788 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28501529
CAS-Nummer: 1017540-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR