6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene 98%

Lieferant: Apollo Scientific

OR9359-1G
APOSOR9359-1GEA 176 EUR
APOSOR9359-1G
6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene 98%
6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene
Formel: C₁₅H₂₁Br
Molecular Weight: 281,231 g/mol
Schmelzpunkt: 92…95 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235072
CAS-Nummer: 119999-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR