4-Isobutyramidobenzeneboronic acid 98%
Lieferant: Apollo Scientific
OR9360-1G
APOSOR9360-1GEA
238
EUR
APOSOR9360-1G
4-Isobutyramidobenzeneboronic acid 98%
4-Isobutyramidobenzeneboronic acid
Formel:
C₁₀H₁₄BNO₃ |
MDL:
MFCD08235068 CAS-Nummer: 874219-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...