4-Methoxy-3-(pyrrolidin-1-ylsulphonyl)benzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Methoxy-3-(pyrrolidin-1-ylsulphonyl)phenyl)boronic acid

OR9362-1G
APOSOR9362-1GEA 315 EUR
APOSOR9362-1G
4-Methoxy-3-(pyrrolidin-1-ylsulphonyl)benzeneboronic acid 95%
4-Methoxy-3-(pyrrolidin-1-ylsulphonyl)benzeneboronic acid
Formel: C₁₁H₁₆BNO₅S
Molecular Weight: 285.124 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08235070
CAS-Nummer: 874219-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR