1,5-Dihydroxynaphthalin 98%

Lieferant: Apollo Scientific

Synonyme: 1,5-Naphthalindiol

OR936200-500G
APOSOR936200-500GEA 55.1 EUR
APOSOR936200-500G
1,5-Dihydroxynaphthalin 98%
1,5-Dihydroxynaphthalin
Formel: C₁₀H₈O₂
Molecular Weight: 160.17 g/mol
Siedepunkt: 375
Schmelzpunkt: 180
Lagertemperatur: Raumtemperatur
MDL: MFCD00003980
CAS-Nummer: 83-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR