1-(1-Cyclohexen-1-yl)piperidine 95%
Lieferant: Apollo Scientific
OR936208-25G
APOSOR936208-25GEA
0
EUR
APOSOR936208-25G
1-(1-Cyclohexen-1-yl)piperidine 95%
1-(1-Cyclohexen-1-yl)piperidine
Formel:
C₁₁H₁₉N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014643 CAS-Nummer: 2981-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...