(2,5-Xylyloxy)essigsäure 95%

Lieferant: Apollo Scientific

Synonyme: (2,5-Dimethylphenoxy)essigsäure

OR936222-25G
APOSOR936222-25GEA 183 EUR
APOSOR936222-25G
(2,5-Xylyloxy)essigsäure 95%
(2,5-Xylyloxy)essigsäure
Formel: (C₆H₃(CH₃)₂)OCH₂COOH
Molecular Weight: 180.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014356
CAS-Nummer: 7356-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR