1-Cyclohexyl-2-pyrrolidinon 95%

Lieferant: Apollo Scientific

OR936238-25G
APOSOR936238-25GEA 161 EUR
APOSOR936238-25G
1-Cyclohexyl-2-pyrrolidinon 95%
1-Cyclohexyl-2-pyrrolidone
Formel: C₁₀H₁₇NO
Molecular Weight: 167,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003191
CAS-Nummer: 6837-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR