2-(Dimethylamino)ethyl-benzoat 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Dimethylamino)ethyl benzoate, Benzoic Acid 2-Dimethylaminoethyl Ester

OR936268-25G
APOSOR936268-25GEA 49.3 EUR
APOSOR936268-25G
2-(Dimethylamino)ethyl-benzoat 95%
2-Dimethylaminoethyl benzoate
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051067
CAS-Nummer: 2208-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR