Methyl-4-acetamido-5-chloro-2-methoxybenzoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-acetamido-5-chloro-2-methoxybenzoate, Metoclopramide Related Compound B

OR936293-100G
APOSOR936293-100GEA 78.3 EUR
APOSOR936293-100G
Methyl-4-acetamido-5-chloro-2-methoxybenzoate 98%
Methyl-4-acetamido-5-chloro-2-methoxybenzoate
Formel: C₁₁H₁₂ClNO₄
Schmelzpunkt: 153…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00048191
CAS-Nummer: 4093-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR