4-Hydroxy-3,5-dimethylbenzoesäure 97%
Lieferant: Apollo Scientific
OR936362-25G
APOSOR936362-25GEA
180
EUR
APOSOR936362-25G
4-Hydroxy-3,5-dimethylbenzoesäure 97%
4-Hydroxy-3,5-dimethylbenzoesäure
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016536 CAS-Nummer: 4919-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...