2-Ethoxy-3-isopropylpyrazin 95%

Lieferant: Apollo Scientific

OR936366-5G
APOSOR936366-5GEA 487 EUR
APOSOR936366-5G
2-Ethoxy-3-isopropylpyrazin 95%
2-Ethoxy-3-isopropylpyrazin
Formel: C₉H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00038029
CAS-Nummer: 72797-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR