trans-4-Methoxy-β-nitrostyrol 96%
Lieferant: Apollo Scientific
Synonyme:
(E)-1-(4-Methoxyphenyl)-2-nitroethene
OR936372-25G
APOSOR936372-25GEA
151
EUR
APOSOR936372-25G
trans-4-Methoxy-β-nitrostyrol 96%
trans-4-Methoxy-β-nitrostyrol
Formel:
C₉H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024826 CAS-Nummer: 5576-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...