N,N-Diethyl-1-naphthalinamin 95%
Lieferant: Apollo Scientific
OR936375-1G
APOSOR936375-1GEA
386
EUR
APOSOR936375-1G
N,N-Diethyl-1-naphthalinamin 95%
N,N-Diethyl-1-naphthalinamin
Formel:
C₁₄H₁₇N Molecular Weight: 199,30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046389 CAS-Nummer: 84-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...