Methyl-5-chloro-5-oxovalerate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl glutaryl chloride, Methyl 4-chloroformylbutyrate

OR936413-25G
APOSOR936413-25GEA 139 EUR
APOSOR936413-25G
Methyl-5-chloro-5-oxovalerate 95%
Methyl-5-chloro-5-oxovalerate
Formel: C₆H₉ClO₃
Molecular Weight: 164.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000756
CAS-Nummer: 1501-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR