trans-4-(2-Nitroethenyl)benzonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
trans-4-(2-Nitrovinyl)benzonitrile, (E)-4-(2-Nitrovinyl)benzonitrile
OR936417-1G
APOSOR936417-1GEA
43.5
EUR
APOSOR936417-1G
trans-4-(2-Nitroethenyl)benzonitrile 95%
trans-4-(2-Nitroethenyl)benzonitrile
Formel:
C₉H₆N₂O₂ Molecular Weight: 174,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00176174 CAS-Nummer: 5153-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...