Dimethylketene methyl trimethylsilyl acetal 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, ((1-Methoxy-2-methylprop-1-en-1-yl)oxy)trimethylsilane

OR936451-25G
APOSOR936451-25GEA 95.7 EUR
APOSOR936451-25G
Dimethylketene methyl trimethylsilyl acetal 95%
Dimethylketene methyl trimethylsilyl acetal
Formel: C₈H₁₈O₂Si
Molecular Weight: 174.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010232
CAS-Nummer: 31469-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR