1,3-Diethyl-2-thioharnstoff 95%

Lieferant: Apollo Scientific

Synonyme: N,N'-Diethylthiourea

OR936470-100G
APOSOR936470-100GEA 125 EUR
APOSOR936470-100G
1,3-Diethyl-2-thioharnstoff 95%
1,3-Diethyl-2-thioharnstoff
Formel: C₅H₁₂N₂S
Molecular Weight: 132,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004925
CAS-Nummer: 105-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR