N-tert-Butyl-3-borono-4-methylbenzolsulfonamid 95%

Lieferant: Apollo Scientific

Synonyme: 5-tert-Butylsulphamoyl-2-methylbenzeneboronic acid

OR9365-250MG
APOSOR9365-250MGEA 288 EUR
APOSOR9365-250MG
N-tert-Butyl-3-borono-4-methylbenzolsulfonamid 95%
N-tert-Butyl-3-borono-4-methylbenzolsulfonamid
Formel: C₁₁H₁₈BNO₄S
Molecular Weight: 271,14 g/mol
Schmelzpunkt: 142…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235063
CAS-Nummer: 874219-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR