1,1-Diphenylhydrazin 95%
Lieferant: Apollo Scientific
OR936542-10G
OR936542-1G
OR936542-5G
APOSOR936542-10GEA
799
EUR
APOSOR936542-10G
APOSOR936542-1G
APOSOR936542-5G
1,1-Diphenylhydrazin 95%
1,1-Diphenylhydrazine
Formel:
C₁₂H₁₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044505 CAS-Nummer: 530-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...