4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) 98%

Lieferant: Apollo Scientific

OR936587-25G OR936587-5G
APOSOR936587-25GEA 522 EUR
APOSOR936587-25G APOSOR936587-5G
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) 98%
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₈H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01734677
CAS-Nummer: 13380-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR